siRNA Details
Gene Symbol: Atp5f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK002960.1 | - | 4 | 373 | ||
AK011312.1 | - | 4 | 387 | |||
AK019459.1 | - | 4 | 391 | |||
AK037892.1 | - | 3 | 2105 | |||
AK149176.1 | - | 4 | 359 | |||
AK150730.1 | - | 5 | 571 | |||
AK151250.1 | - | 5 | 571 | |||
AK151574.1 | - | 4 | 364 | |||
AK152253.1 | - | 4 | 391 | |||
AK152829.1 | - | 4 | 391 | |||
AK157544.1 | - | 4 | 427 | |||
AK158569.1 | - | 4 | 414 | |||
AK162311.1 | - | 4 | 356 | |||
AK167480.1 | - | 4 | 393 | |||
AK167626.1 | - | 4 | 364 | |||
AK168028.1 | - | 4 | 389 | |||
AK168041.1 | - | 4 | 390 | |||
AK168590.1 | - | 4 | 364 | |||
AK169137.1 | - | 4 | 391 | |||
AK169634.1 | - | 4 | 346 | |||
BC049640.1 | - | 4 | 321 | |||
BC087882.1 | - | 6 | 875 | 6706159 | ||
BC089574.1 | - | 4 | 353 | 6756180 |

More Information
Other Species Targeted

