siRNA Details
Gene Symbol: Atp5j | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA259963.1 | - | 3, 4 | 335 | ||
AK078484.1 | - | 3, 4 | 496 | |||
BC010766.1 | - | 4, 5 | 631 | 4219715 | ||
BE851144.1 | - | 4, 5 | 373 | |||
U77128.1 | - | 2, 3 | 294 | |||
W75852.1 | - | 3, 4 | 361 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
