siRNA Details
Gene Symbol: Atp5j | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA259963.1 | - | 3 | 273 | ||
AK078484.1 | - | 3 | 434 | |||
BC010766.1 | - | 4 | 569 | 4219715 | ||
BE851144.1 | - | 4 | 311 | |||
BY079469.1 | - | 3 | 348 | |||
U77128.1 | - | 2 | 232 | |||
W75852.1 | - | 3 | 299 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
