siRNA Details
Gene Symbol: Atp5k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007507.2 | NP_031533.2 | 3 | 150 | ||
GenBank mRNA | BC028438.1 | - | 3 | 150 | 1332765 | |
BC089550.1 | - | 3 | 142 | 30380642 | ||
S52977.1 | - | 3 | 133 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
