siRNA Details
Gene Symbol: Atp6v1b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007509.3 | NP_031535.2 | 13, 14 | 1460 | ||
GenBank mRNA | AK146499.1 | - | 13, 14 | 1425 | ||
AK151200.1 | - | 13, 14 | 1461 | |||
AK151322.1 | - | 13, 14 | 1433 | |||
AK151586.1 | - | 13, 14 | 1433 | |||
AK152718.1 | - | 13, 14 | 1433 | |||
AK152766.1 | - | 13, 14 | 1433 | |||
AK153341.1 | - | 13, 14 | 1424 | |||
AK159133.1 | - | 13, 14 | 1431 | |||
AK159153.1 | - | 13, 14 | 1431 | |||
AK159586.1 | - | 13, 14 | 1428 | |||
AK159701.1 | - | 13, 14 | 1432 | |||
AK159986.1 | - | 13, 14 | 1432 | |||
AK160080.1 | - | 13, 14 | 1431 | |||
AK160854.1 | - | 13, 14 | 1426 | |||
AK166669.1 | - | 13, 14 | 1400 | |||
AK168852.1 | - | 13, 14 | 1424 | |||
AK169155.1 | - | 13, 14 | 1439 | |||
AK169270.1 | - | 13, 14 | 1436 | |||
AK170110.1 | - | 13, 14 | 1455 | |||
AK187518.1 | - | 1, 2 | 111 | |||
AK205553.1 | - | 1, 2 | 111 | |||
BC012497.1 | - | 13, 14 | 1425 | 4500843 | ||
BC046302.1 | - | 13, 14 | 1415 | 6492612 | ||
BC085300.1 | - | 13, 14 | 1407 | 6305977 | ||
EF437356.1 | - | 3, 4 | 291 | |||
Y12634.1 | - | 13, 14 | 1417 |
