siRNA Details
Gene Symbol: Atp6v1b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007509.3 | NP_031535.2 | 10, 11 | 1136 | ||
GenBank mRNA | AK146499.1 | - | 10, 11 | 1101 | ||
AK151200.1 | - | 10, 11 | 1137 | |||
AK151322.1 | - | 10, 11 | 1109 | |||
AK151586.1 | - | 10, 11 | 1109 | |||
AK152718.1 | - | 10, 11 | 1109 | |||
AK152766.1 | - | 10, 11 | 1109 | |||
AK153341.1 | - | 10, 11 | 1100 | |||
AK159133.1 | - | 10, 11 | 1107 | |||
AK159153.1 | - | 10, 11 | 1107 | |||
AK159586.1 | - | 10, 11 | 1104 | |||
AK159701.1 | - | 10, 11 | 1108 | |||
AK159986.1 | - | 10, 11 | 1108 | |||
AK160080.1 | - | 10, 11 | 1107 | |||
AK160854.1 | - | 10, 11 | 1102 | |||
AK166669.1 | - | 10, 11 | 1076 | |||
AK168852.1 | - | 10, 11 | 1100 | |||
AK169155.1 | - | 10, 11 | 1115 | |||
AK169270.1 | - | 10, 11 | 1112 | |||
AK170110.1 | - | 10, 11 | 1131 | |||
AK185178.1 | - | 1, 2 | 66 | |||
AK189307.1 | - | 1, 2 | 71 | |||
AK190992.1 | - | 1, 2 | 71 | |||
AK196704.1 | - | 1, 2 | 71 | |||
AK198404.1 | - | 1, 2 | 72 | |||
AK216659.1 | - | 1, 2 | 71 | |||
AK218365.1 | - | 1, 2 | 71 | |||
BC012497.1 | - | 10, 11 | 1101 | 4500843 | ||
BC046302.1 | - | 10, 11 | 1091 | 6492612 | ||
BC085300.1 | - | 10, 11 | 1083 | 6305977 | ||
Y12634.1 | - | 10, 11 | 1093 |
