siRNA Details
Gene Symbol: Atp6v0e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025272.2 | NP_079548.1 | 2 | 207 | ||
GenBank mRNA | AK007610.1 | - | 2 | 186 | ||
AK008464.1 | - | 2 | 187 | |||
AK013229.1 | - | 2 | 168 | |||
AK017760.1 | - | 2 | 209 | |||
AK030335.1 | - | 2 | 159 | |||
AK134836.1 | - | 2 | 191 | |||
AK159669.1 | - | 2 | 195 | |||
AK168015.1 | - | 2 | 176 | |||
BC024733.1 | - | 2 | 171 | 5374017 | ||
BY707658.1 | - | 2 | 183 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
