siRNA Details
Gene Symbol: Atpif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF002718.1 | - | 2, 3 | 212 | ||
AK008346.1 | - | 2, 3 | 210 | |||
AK152389.1 | - | 3, 4 | 1227 | |||
AV462016.1 | - | 2, 3 | 273 | |||
BC012680.1 | - | 2, 3 | 271 | 4217188 | ||
BU055998.1 | - | 2, 3 | 198 | |||
CB848021.2 | - | 2, 3 | 235 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
