siRNA Details
Gene Symbol: Atp6v0c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009729.3 | NP_033859.1 | 3 | 530 | ||
GenBank mRNA | AK002570.1 | - | 3 | 539 | ||
AK002871.1 | - | 3 | 518 | |||
AK028239.1 | - | 3 | 520 | |||
AK146159.1 | - | 3 | 496 | |||
AK170346.1 | - | 3 | 496 | |||
BC050939.1 | - | 3 | 480 | |||
BC057084.1 | - | 3 | 1947 | |||
BC083129.1 | - | 3 | 510 | 6440462 | ||
BC099475.1 | - | 3 | 473 | 30938468 | ||
BC132267.1 | - | 3 | 419 | 40130544 | ||
BC132611.1 | - | 3 | 419 | 40130888 | ||
M64298.1 | - | 3 | 532 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
