siRNA Details
Gene Symbol: Slc7a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044740.2 | NP_001038205.1 | 5 | 918 | ||
NM_007514.3 | NP_031540.2 | 5 | 918 | |||
GenBank mRNA | AK154621.1 | - | 5 | 892 | ||
AK155558.1 | - | 5 | 893 | |||
AK171369.1 | - | 5 | 890 | |||
BC021653.1 | - | 5 | 913 | |||
BC127082.1 | - | 5 | 834 | 40111372 | ||
DQ086834.1 | - | 5 | 873 | |||
L03290.1 | - | 4 | 720 | |||
L11600.1 | - | 4 | 720 | |||
L29006.1 | - | 4 | 1243 | |||
M62838.1 | - | 5 | 831 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
