siRNA Details
Gene Symbol: Slc7a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044740.2 | NP_001038205.1 | 2 | 500 | ||
NM_007514.3 | NP_031540.2 | 2 | 500 | |||
GenBank mRNA | AK154621.1 | - | 2 | 474 | ||
AK155558.1 | - | 2 | 475 | |||
AK171369.1 | - | 2 | 472 | |||
BB648063.1 | - | 2 | 474 | |||
BC021653.1 | - | 2 | 494 | |||
BC127082.1 | - | 2 | 416 | 40111372 | ||
CN710433.1 | - | 1 | 107 | |||
DQ086834.1 | - | 2 | 455 | |||
L03290.1 | - | 1 | 302 | |||
L11600.1 | - | 1 | 302 | |||
L29006.1 | - | 1 | 825 | |||
M62838.1 | - | 2 | 413 |

More Information
Other Species Targeted

Gene Ontology Categories:
