siRNA Details
Gene Symbol: Hnrnpd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077265.2 | NP_001070733.1 | 4 | 813 | ||
NM_001077266.2 | NP_001070734.1 | 3 | 756 | |||
NM_001077267.2 | NP_001070735.1 | 3 | 756 | |||
NM_007516.3 | NP_031542.2 | 4 | 813 | |||
GenBank mRNA | AK077409.1 | - | 4 | 802 | ||
AK135733.1 | - | 4 | 759 | |||
BC011172.1 | - | 4 | 488 | |||
BC049098.1 | - | 4 | 813 | 5717286 | ||
CA451481.1 | - | 2 | 94 | |||
CK618758.1 | - | 3 | 382 | |||
U11273.1 | - | 3 | 593 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
