siRNA Details
Gene Symbol: Auh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016709.2 | NP_057918.2 | 2, 3 | 289 | ||
GenBank mRNA | AK003219.1 | - | 2, 3 | 268 | ||
AK003929.1 | - | 2, 3 | 269 | |||
AK019978.1 | - | 2, 3 | 266 | |||
AK134886.1 | - | 2, 3 | 261 | |||
AK147855.1 | - | 2, 3 | 293 | |||
AK169661.1 | - | 2, 3 | 267 | |||
BC026525.1 | - | 2, 3 | 227 | |||
BC049597.1 | - | 2, 3 | 273 | 6593786 | ||
BY741014.1 | - | 2, 3 | 290 |
