siRNA Details
Gene Symbol: Auh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016709.2 | NP_057918.2 | 3 | 346 | ||
GenBank mRNA | AK003219.1 | - | 3 | 325 | ||
AK003929.1 | - | 3 | 326 | |||
AK019978.1 | - | 3 | 323 | |||
AK134886.1 | - | 3 | 318 | |||
AK147855.1 | - | 3 | 350 | |||
AK169661.1 | - | 3 | 324 | |||
BC026525.1 | - | 3 | 284 | |||
BC049597.1 | - | 3 | 330 | 6593786 | ||
BY741014.1 | - | 3 | 347 |
