siRNA Details
Gene Symbol: Aup1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007517.4 | NP_031543.2 | 3 | 543 | ||
GenBank mRNA | AK144497.1 | - | 1 | 495 | ||
AK154707.1 | - | 3 | 412 | |||
AK158035.1 | - | 3 | 334 | |||
AK167335.1 | - | 3 | 333 | |||
AK171011.1 | - | 3 | 346 | |||
AK171184.1 | - | 3 | 338 | |||
AK171202.1 | - | 3 | 352 | |||
AK171249.1 | - | 3 | 332 | |||
AK171541.1 | - | 3 | 333 | |||
AK171621.1 | - | 3 | 325 | |||
BC016485.1 | - | 3 | 336 | 3962982 | ||
U41736.1 | - | 3 | 341 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
