siRNA Details
Gene Symbol: Axin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015732.4 | NP_056547.3 | 2 | 764 | ||
GenBank mRNA | AF073788.1 | - | 2 | 629 | ||
AF205889.1 | - | 1 | 469 | |||
AK049192.1 | - | 2 | 708 | |||
AK142333.1 | - | 2 | 766 | |||
BC057338.1 | - | 2 | 747 | 6827741 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
