siRNA Details
Gene Symbol: Bcap29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164090.1 | NP_001157562.1 | 4 | 300 | ||
NM_007530.3 | NP_031556.1 | 3 | 463 | |||
GenBank mRNA | AK090068.1 | - | 4 | 277 | ||
AK144754.1 | - | 4 | 274 | |||
AK218818.1 | - | 3 | 201 | |||
BC003303.1 | - | 3 | 367 | 3584161 | ||
BC021661.1 | - | 3 | 474 | 2646883 | ||
X78684.1 | - | 3 | 275 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
