siRNA Details
Gene Symbol: Phb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007531.2 | NP_031557.2 | 8 | 960 | ||
GenBank mRNA | AK002334.1 | - | 8 | 926 | ||
AK011107.1 | - | 8 | 927 | |||
AK090088.1 | - | 8 | 917 | |||
AK132052.1 | - | 8 | 922 | |||
AK132058.1 | - | 8 | 949 | |||
AK135729.1 | - | 8 | 877 | |||
AK151952.1 | - | 8 | 954 | |||
AK152375.1 | - | 7 | 1609 | |||
AK153368.1 | - | 7 | 1609 | |||
AY211613.1 | - | 8 | 922 | |||
AY319418.1 | - | 8 | 812 | |||
BC145875.1 | - | 8 | 912 | 40131178 | ||
X78683.1 | - | 8 | 916 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
