siRNA Details
Gene Symbol: Bcl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009741.4 | NP_033871.2 | 1 | 1930 | ||
NM_177410.2 | NP_803129.2 | 1 | 1930 | |||
GenBank mRNA | AK049473.1 | - | 1 | 1924 | ||
AK077913.1 | - | 1 | 710 | |||
BC068988.1 | - | 1 | 818 | 1265430 | ||
BC095964.1 | - | 1 | 1586 | 6306635 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: AF512835.
Gene Ontology Categories:
