siRNA Details
Gene Symbol: Bcl2a1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009742.3 | NP_033872.1 | 2 | 745 | ||
GenBank mRNA | BC028762.1 | - | 691 | 1226745 | ||
L16462.1 | - | 2 | 745 |
Gene Symbol: Bcl2a1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007534.3 | NP_031560.2 | 2 | 701 | ||
GenBank mRNA | AK138369.1 | - | 2 | 681 | ||
AK152200.1 | - | 2 | 671 | |||
BC100462.1 | - | 2 | 678 | 30295442 | ||
BP773251.1 | - | 2 | 259 |
Gene Symbol: Bcl2a1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007536.2 | NP_031562.1 | 2 | 745 | ||
GenBank mRNA | AK165576.1 | - | 2 | 497 | ||
BC027536.1 | - | 2 | 658 | 1363928 | ||
BC053965.1 | - | 2 | 767 |

More Information
Other Species Targeted

Gene Ontology Categories:
