siRNA Details
Gene Symbol: Bcl2a1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007536.2 | NP_031562.1 | 2 | 749 | ||
GenBank mRNA | AK165576.1 | - | 2 | 501 | ||
BC027536.1 | - | 2 | 662 | 1363928 | ||
BC053965.1 | - | 2 | 771 |
Gene Symbol: Bcl2a1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009742.3 | NP_033872.1 | 2 | 749 | ||
GenBank mRNA | BC028762.1 | - | 695 | 1226745 | ||
L16462.1 | - | 2 | 749 |
Gene Symbol: Bcl2a1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007534.3 | NP_031560.2 | 2 | 705 | ||
GenBank mRNA | AK138369.1 | - | 2 | 685 | ||
AK152200.1 | - | 2 | 675 | |||
BC100462.1 | - | 2 | 682 | 30295442 | ||
BP773251.1 | - | 2 | 263 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
