siRNA Details
Gene Symbol: Bid | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007544.3 | NP_031570.2 | 5 | 686 | ||
GenBank mRNA | AK045731.1 | - | 5 | 701 | ||
AK051076.1 | - | 702 | ||||
AK052356.1 | - | 5 | 691 | |||
AK077657.1 | - | 5 | 669 | |||
AK161235.1 | - | 5 | 708 | |||
BC002031.1 | - | 5 | 686 | 3486537 | ||
U75506.1 | - | 4 | 563 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
