siRNA Details
Gene Symbol: Cxcr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007551.2 | NP_031577.2 | 1, 2 | 92 | ||
GenBank mRNA | AK089611.1 | - | 1, 2 | 105 | ||
AK133064.1 | - | 3, 4 | 324 | |||
AK155954.1 | - | 1, 2 | 89 | |||
AK156087.1 | - | 1, 2 | 89 | |||
AK157016.1 | - | 1, 2 | 89 | |||
BC064059.1 | - | 1, 2 | 92 | 1366773 | ||
X71788.1 | - | 1, 2 | 49 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
