siRNA Details
Gene Symbol: Bmp15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009757.4 | NP_033887.1 | 2 | 900 | ||
GenBank mRNA | AF082348.1 | - | 2 | 783 | ||
AJ010259.1 | - | 1 | 127 | |||
AJ132406.1 | - | 2 | 540 | |||
AK028735.1 | - | 2 | 586 | |||
AK135757.1 | - | 2 | 807 | |||
AK135882.1 | - | 2 | 903 | |||
AK136127.1 | - | 2 | 294 | |||
AK136129.1 | - | 2 | 294 | |||
AK139773.1 | - | 2 | 901 | |||
AK139847.1 | - | 2 | 902 | |||
AK139936.1 | - | 2 | 856 | |||
BC055363.1 | - | 2 | 897 | 30051425 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
