siRNA Details
Gene Symbol: Bmpr1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009758.4 | NP_033888.2 | 11, 12 | 1757 | ||
GenBank mRNA | AK132051.1 | - | 11, 12 | 1770 | ||
AK143804.1 | - | 2, 3 | 384 | |||
AY365062.1 | - | 9, 10 | 1351 | |||
BC042611.1 | - | 10, 11 | 1547 | 5364272 | ||
D16250.1 | - | 9, 10 | 1492 | |||
U04672.1 | - | 9, 10 | 1361 | |||
Z23154.1 | - | 9, 10 | 1351 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
