siRNA Details
Gene Symbol: Bnc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007562.2 | NP_031588.2 | 5 | 2827 | ||
GenBank mRNA | AK087567.1 | - | 5 | 2822 | ||
BC127226.1 | - | 1 | 494 | |||
U88064.1 | - | 5 | 2774 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
