siRNA Details
Gene Symbol: Bop1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013481.1 | NP_038509.1 | 6 | 754 | ||
GenBank mRNA | AK079627.1 | - | 6 | 734 | ||
AK144095.1 | - | 6 | 765 | |||
AK167110.1 | - | 6 | 737 | |||
AK167225.1 | - | 6 | 738 | |||
AK167829.1 | - | 6 | 732 | |||
AK167878.1 | - | 6 | 742 | |||
AK168127.1 | - | 6 | 735 | |||
BC012693.1 | - | 6 | 728 | 4193123 | ||
U77415.1 | - | 6 | 754 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
