siRNA Details
Gene Symbol: Brca1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009764.3 | NP_033894.3 | 10 | 3164 | ||
GenBank mRNA | AB221610.1 | - | 1 | 393 | ||
BC068303.1 | - | 10 | 3000 | 30431022 | ||
U31625.1 | - | 10 | 3021 | |||
U32446.1 | - | 10 | 3074 | |||
U35641.1 | - | 10 | 3057 | |||
U36475.1 | - | 9 | 2984 | |||
U68174.1 | - | 10 | 3068 |
