siRNA Details
Gene Symbol: Birc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007566.3 | NP_031592.3 | 3 | 730 | ||
GenBank mRNA | AK045446.1 | - | 3 | 499 | ||
AK079995.1 | - | 3 | 507 | |||
Y17267.1 | - | 3 | 731 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Birc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007566.3 | NP_031592.3 | 3 | 730 | ||
GenBank mRNA | AK045446.1 | - | 3 | 499 | ||
AK079995.1 | - | 3 | 507 | |||
Y17267.1 | - | 3 | 731 |