siRNA Details
Gene Symbol: Btg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007569.2 | NP_031595.1 | 2 | 723 | ||
GenBank mRNA | AK149547.1 | - | 2 | 717 | ||
AK162764.1 | - | 2 | 719 | |||
BC006834.1 | - | 2 | 697 | 3597853 | ||
BC018309.1 | - | 2 | 709 | 4190018 | ||
L16846.1 | - | 2 | 714 |

More Information
Other Species Targeted

Gene Ontology Categories:
