siRNA Details
Gene Symbol: Btk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013482.2 | NP_038510.2 | 16 | 1742 | ||
GenBank mRNA | BC053392.1 | - | 16 | 1678 | 30075322 | |
L08967.1 | - | 16 | 1718 | |||
L10627.1 | - | 16 | 1750 | |||
L29788.1 | - | 16 | 1714 |

More Information
Other Species Targeted

Gene Ontology Categories:
