siRNA Details
Gene Symbol: C1qa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007572.2 | NP_031598.2 | 3 | 751 | ||
GenBank mRNA | AK002655.1 | - | 3 | 677 | ||
AK136737.1 | - | 3 | 677 | |||
AK150771.1 | - | 3 | 677 | |||
AK159210.1 | - | 3 | 677 | |||
AK159343.1 | - | 3 | 672 | |||
BC002086.1 | - | 3 | 710 | 3592169 | ||
BC020096.1 | - | 2 | 672 | |||
BY698724.1 | - | 1 | 62 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
