siRNA Details
Gene Symbol: C1qb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009777.2 | NP_033907.1 | 3 | 589 | ||
GenBank mRNA | AK140712.1 | - | 3 | 546 | ||
AK152764.1 | - | 3 | 608 | |||
AK153124.1 | - | 3 | 481 | |||
BC067001.1 | - | 3 | 525 | 5715633 | ||
M22531.1 | - | 3 | 589 | |||
M36293.1 | - | 3 | 589 | |||
X16874.1 | - | 2 | 444 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
