siRNA Details
Gene Symbol: Cacna1e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009782.3 | NP_033912.2 | 1 | 215 | ||
GenBank mRNA | AK039991.1 | - | 3 | 739 | ||
AK171983.1 | - | 2 | 773 | |||
BC150750.1 | - | 1 | 326 | 9087661 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_019294, XM_006250069, L15453, AY029412.
Gene Ontology Categories:
