siRNA Details
Gene Symbol: Cacnb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037099.2 | NP_001032176.1 | 11 | 1270 | ||
NM_001285426.1 | NP_001272355.1 | 11 | 1054 | |||
NM_001285427.1 | NP_001272356.1 | 11 | 1350 | |||
NM_001285428.1 | NP_001272357.1 | 10 | 979 | |||
NM_146123.3 | NP_666235.1 | 10 | 1190 | |||
GenBank mRNA | AF039417.1 | - | 1 | 47 | ||
AK032311.1 | - | 11 | 1493 | |||
AK038633.1 | - | 11 | 1306 | |||
AK043850.1 | - | 10 | 1133 | |||
AK079616.1 | - | 10 | 1140 | |||
AK147338.1 | - | 11 | 1244 | |||
AK158606.1 | - | 10 | 981 | |||
BC026479.1 | - | 10 | 976 | 4501980 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
