siRNA Details
Gene Symbol: Ddr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198831.1 | NP_001185760.1 | 6 | 721 | ||
NM_001198833.1 | NP_001185762.1 | 6 | 840 | |||
NM_007584.2 | NP_031610.1 | 6 | 896 | |||
NM_172962.1 | NP_766550.1 | 6 | 896 | |||
GenBank mRNA | AF026259.1 | - | 6 | 738 | ||
AK031442.1 | - | 6 | 799 | |||
AK138698.1 | - | 6 | 708 | |||
AK143088.1 | - | 6 | 789 | |||
BC037108.1 | - | 7 | 975 | |||
BC065998.1 | - | 6 | 732 | 5686085 | ||
L57509.1 | - | 6 | 744 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
