siRNA Details
Gene Symbol: Aspm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009791.4 | NP_033921.3 | 6 | 2449 | ||
GenBank mRNA | AF533752.1 | - | 6 | 2427 | ||
AK052326.1 | - | 6 | 2450 | |||
AY971958.1 | - | 6 | 2290 |

More Information
Other Species Targeted

Gene Ontology Categories:
