siRNA Details
Gene Symbol: Aspm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009791.4 | NP_033921.3 | 18 | 4465 | ||
GenBank mRNA | AF533752.1 | - | 18 | 4443 | ||
BC026699.1 | - | 2 | 516 | |||
BI989704.1 | - | 2 | 463 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
