siRNA Details
Gene Symbol: Camk2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286809.1 | NP_001273738.1 | 5 | 723 | ||
NM_009792.3 | NP_033922.1 | 6 | 756 | |||
NM_177407.4 | NP_803126.1 | 17 | 1444 | |||
GenBank mRNA | AK083245.1 | - | 17 | 1445 | ||
AK122410.2 | - | 17 | 1440 | |||
BC031745.1 | - | 17 | 1425 | 4506005 | ||
X14836.1 | - | 17 | 1428 | |||
X87142.1 | - | 6 | 756 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
