siRNA Details
Gene Symbol: Capza3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007605.4 | NP_031631.3 | 1 | 466 | ||
GenBank mRNA | AK016391.1 | - | 1 | 435 | ||
BC049620.1 | - | 1 | 434 | 6770504 | ||
BY707143.1 | - | 1 | 466 | |||
D87471.1 | - | 1 | 424 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
