siRNA Details
Gene Symbol: Car6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009802.2 | NP_033932.2 | 4, 5 | 519 | ||
GenBank mRNA | AF079834.1 | - | 3, 4 | 609 | ||
AF079835.1 | - | 4, 5 | 546 | |||
AK132599.1 | - | 4, 5 | 520 | |||
AK161216.1 | - | 4, 5 | 515 | |||
BC006726.1 | - | 4, 5 | 606 | |||
BC018582.1 | - | 4, 5 | 496 | |||
BC036270.1 | - | 4, 5 | 502 | |||
BC039657.1 | - | 4, 5 | 504 | |||
BC046495.1 | - | 3, 4 | 295 | |||
BC049973.1 | - | 4, 5 | 517 | |||
BC055437.1 | - | 4, 5 | 505 | 4190566 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
