siRNA Details
Gene Symbol: Car7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301164.1 | NP_001288093.1 | 7 | 1201 | ||
NM_001301165.1 | NP_001288094.1 | 7 | 769 | |||
NM_053070.3 | NP_444300.1 | 7 | 796 | |||
GenBank mRNA | AF291660.1 | - | 6 | 691 | ||
AK090390.1 | - | 7 | 800 | |||
AK144275.1 | - | 7 | 770 | |||
AY075021.1 | - | 7 | 797 | |||
BC038270.1 | - | 7 | 1176 | |||
BC094913.1 | - | 7 | 784 | 30016726 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
