siRNA Details
Gene Symbol: Casp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284409.1 | NP_001271338.1 | 5 | 656 | ||
NM_009810.3 | NP_033940.1 | 5 | 651 | |||
GenBank mRNA | AK080651.1 | - | 2 | 166 | ||
AK146226.1 | - | 5 | 657 | |||
BC038825.1 | - | 5 | 540 | 4482368 | ||
D86352.1 | - | 5 | 470 | |||
U19522.1 | - | 5 | 467 | |||
U49929.1 | - | 5 | 493 | |||
U63720.1 | - | 2 | 259 | |||
Y13086.1 | - | 5 | 497 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
