siRNA Details
Gene Symbol: Casp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284409.1 | NP_001271338.1 | 4, 5 | 525 | ||
NM_009810.3 | NP_033940.1 | 4, 5 | 520 | |||
GenBank mRNA | AK080651.1 | - | 1, 2 | 35 | ||
AK146226.1 | - | 4, 5 | 526 | |||
BC038825.1 | - | 4, 5 | 409 | 4482368 | ||
D86352.1 | - | 4, 5 | 339 | |||
U19522.1 | - | 4, 5 | 336 | |||
U49929.1 | - | 4, 5 | 362 | |||
U63720.1 | - | 1, 2 | 128 | |||
Y13086.1 | - | 4, 5 | 366 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
