siRNA Details
Gene Symbol: Casp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007611.2 | NP_031637.1 | 6, 7 | 879 | ||
GenBank mRNA | AK154978.1 | - | 6, 7 | 759 | ||
BC005428.1 | - | 6, 7 | 736 | 3600155 | ||
CT010348.1 | - | 5, 6 | 690 | |||
D86353.1 | - | 6, 7 | 742 | |||
Y13088.1 | - | 6, 7 | 879 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | U67321.1 | - | 1274 | Chr. - - [-] on Build |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
