siRNA Details
Gene Symbol: Casr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013803.2 | NP_038831.2 | 3 | 811 | ||
GenBank mRNA | AF110178.1 | - | 3 | 812 | ||
AF110179.1 | - | 3 | 812 | |||
AF128842.1 | - | 2 | 509 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
