siRNA Details
Gene Symbol: Cav3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007617.3 | NP_031643.1 | 2 | 503 | ||
GenBank mRNA | BC024383.1 | - | 2 | 513 | 4988598 | |
CJ057698.1 | - | 2 | 280 | |||
U36579.1 | - | 2 | 317 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
