siRNA Details
Gene Symbol: Serpina6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007618.2 | NP_031644.1 | 4 | 953 | ||
GenBank mRNA | AK145842.1 | - | 4 | 985 | ||
BC013632.1 | - | 4 | 972 | 4194164 | ||
X70533.1 | - | 4 | 953 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
