siRNA Details
Gene Symbol: GUCY1A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000856.5 | NP_000847.2 | 7 | 1216 | ||
NM_001130682.2 | NP_001124154.1 | 6 | 1122 | |||
NM_001130683.3 | NP_001124155.1 | 6 | 1090 | |||
NM_001130684.2 | NP_001124156.1 | 5 | 1062 | |||
NM_001130685.2 | NP_001124157.1 | 6 | 911 | |||
NM_001130687.2 | NP_001124159.1 | 6 | 1122 | |||
NM_001256449.1 | NP_001243378.1 | 6 | 1053 | |||
XM_005262955.2 | XP_005263012.1 | 6 | 1084 | |||
XM_005262956.2 | XP_005263013.1 | 7 | 899 | |||
XM_005262957.2 | XP_005263014.1 | 7 | 1004 | |||
XM_006714196.2 | XP_006714259.1 | 6 | 1071 | |||
XM_006714197.2 | XP_006714260.1 | 6 | 846 | |||
XM_006714198.2 | XP_006714261.1 | 6 | 731 | |||
XM_011531900.2 | XP_011530202.1 | 6 | 810 | |||
GenBank mRNA | AK096588.1 | - | 6 | 1053 | ||
AK226125.1 | - | 6 | 779 | |||
AK309950.1 | - | 6 | 984 | |||
AK312952.1 | - | 6 | 1022 | |||
BC028384.2 | - | 5 | 1062 | 4792661 | ||
BC095395.1 | - | 6 | 1013 | |||
BX647200.1 | - | 6 | 889 | |||
BX649180.1 | - | 6 | 843 | |||
U58855.1 | - | 4 | 697 | |||
X66534.1 | - | 7 | 1196 | |||
Y15723.1 | - | 7 | 1203 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
