siRNA Details
Gene Symbol: Cbx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007625.2 | NP_031651.2 | 5 | 1006 | ||
GenBank mRNA | AK155152.1 | - | 1 | 104 | ||
AK155512.1 | - | 1 | 104 | |||
BC018304.1 | - | 5 | 1037 | |||
BC115843.1 | - | 2 | 683 | |||
BC115844.1 | - | 1 | 898 | |||
BC117801.1 | - | 5 | 880 | 40092282 | ||
BC117802.1 | - | 5 | 911 | 40092283 | ||
U63387.1 | - | 5 | 918 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
